EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Nucleus accumbens-derived glial cell line-derived neurotrophic factor is a retrograde enhancer of dopaminergic tone in the mesocorticolimbic system.
PMID 20980608
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-257
Anti-p300 CT Antibody, clone RW128, clone RW128, Upstate®, from mouse