EMAIL THIS PAGE TO A FRIEND
The Journal of bone and joint surgery. American volume
Arthroplasty of the temporomandibular joint in children with interposition of tantalum foil.
PMID 20992205
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta
GF32441543
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.99%
Ta
GF48780142
Tantalum, foil, 100x100mm, thickness 0.5mm, annealed, 99.9%
Ta
GF66145260
Tantalum, foil, 100x100mm, thickness 0.5mm, annealed, polished on both sides, 99.9%
Ta
GF55289548
Tantalum, foil, 15mm disks, thickness 0.5mm, annealed, 99.9%
Ta
GF64539872
Tantalum, foil, 15mm disks, thickness 0.75mm, annealed, 99.9%
Ta
GF24066525
Tantalum, foil, 1m coil, thickness 0.006mm, 99.9%
Ta
GF50723243
Tantalum, foil, 1m coil, thickness 0.009mm, 99.9%
Ta
GF45003204
Tantalum, foil, 1m coil, thickness 0.015mm, 99.9%
Ta
GF94153754
Tantalum, foil, 1m coil, thickness 0.025mm, as rolled, 99.9%
Ta
GF19196310
Tantalum, foil, 1m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF13236487
Tantalum, foil, 1m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53856186
Tantalum, foil, 1m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF10672365
Tantalum, foil, 1m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF67523351
Tantalum, foil, 1m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta