EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Synthesis of substituted aminobenzacridines.
PMID 20994994
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
384003
2-Bromo-5-methoxybenzoic acid, 98%
C8H7BrO3