EMAIL THIS PAGE TO A FRIEND
Drugs under experimental and clinical research
Ramoplanin: a review of its discovery and its chemistry.
PMID 2100246
Abstract
Ramoplanin is a novel antibacterial agent with characteristics that make it suitable for development as a topical treatment for acne, infected wounds, and for treatment of antibiotic-associated diarrhoea. In this paper, the authors will review the discovery process and the descriptive chemistry of this novel molecular entity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R1781 Ramoplanin