EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Synthesis and characterization of amphiphilic glycidol-chitosan-deoxycholic acid nanoparticles as a drug carrier for doxorubicin.
PMID 21028798
Abstract
Novel amphiphilic chitosan derivatives (glycidol-chitosan-deoxycholic acid, G-CS-DCA) were synthesized by grafting hydrophobic moieties, deoxycholic acid (DCA), and hydrophilic moieties, glycidol, with the purpose of preparing carriers for poorly soluble drugs. Based on self-assembly, G-CS-DCA can form nanoparticles with size ranging from 160 to 210 nm, and G-CS-DCA nanoparticles maintained stable structure for about 3 months when stored in PBS (pH 7.4) at room temperature. The critical aggregation concentration decreased from 0.043 mg/mL to 0.013 mg/mL with the increase of degree of substitution (DS) of DCA. Doxorubicin (DOX) could be easily encapsulated into G-CS-DCA nanoparticles and keep a sustained release manner without burst release when exposed to PBS (pH 7.4) at 37 °C. Antitumor efficacy results showed that DOX-G-CS-DCA have significant antitumor activity when MCF-7 cells were incubated with different concentration of DOX-G-CS-DCA nanoparticles. The fluorescence imaging results indicated DOX-G-CS-DCA nanoparticles could easily be uptaken by MCF-7 cells. These results suggested that G-CS-DCA nanoparticles may be a promising carrier for DOX delivery in cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480819
(R)-(+)-Glycidol, 97%, optical purity ee: 98% (GLC)
C3H6O2
G5809
Glycidol, 96%
C3H6O2