EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
GMI, a Ganoderma immunomodulatory protein, down-regulates tumor necrosis factor ?-induced expression of matrix metalloproteinase 9 via NF-?B pathway in human alveolar epithelial A549 cells.
PMID 21028821