EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Charge regulation enables anionic hydroxypropyl guar-borate adsorption onto anionic and cationic polystyrene latex.
PMID 21030034
Abstract
Reported are adsorption isotherms for guar and hydroxypropyl guar (HPG), with and without the presence of borate ions, onto surfactant free anionic polystyrene latex. Guar and HPG formed adsorbed monolayers on the hydrophobic latex. The presence of borate ions converted the nonionic guar and HPG into an anionic polyelectrolyte. However, there was no measurable influence of bound borate ions on the adsorption of guar or HPG onto anionic, hydrophobic latex. To underscore the unusual behavior of HPG-borate, a sample of HPG was oxidized to introduce carboxyl groups, and the adsorption of the carboxylated HPG onto anionic polystyrene was measured. Unlike HPG-borate, oxidized HPG did not adsorb onto negative polystyrene latex at neutral pH because of electrostatic repulsion. To explain the adsorption of negative HPG-borate onto negative latex, we proposed that as HPG-borate segments approach the latex surface, the negative electrostatic potential near the latex surface induces the detachment of the labile borate groups from HPG.