EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Alkyne-stabilized ruthenium nanoparticles: manipulation of intraparticle charge delocalization by nanoparticle charge States.
PMID 21031389
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
244465
1-Octyne, 97%
C8H14