EMAIL THIS PAGE TO A FRIEND
Wei sheng yan jiu = Journal of hygiene research
[Effect of diisobutyl phthalate on antioxidase activity and DNA damage in mice].
PMID 21033427
Abstract
To investigate the oxidative damage of mice induced by diisobutyl phthalate (DiBP) and the mechanism of free radical oxidative damage caused by DiBP. Sixty KunMing mice were divided by weight into 5 groups after accommodation to the experimental animal room for 3 days. The control group and 4 DiBP groups, group I, II, III and IV, were given DiBP in corn oil by gavages at the dosage of 0, 50, 250, 500 and 1000 mg/kg respectively. The mice were fed with normal diets and drinking water freely for 8 weeks. By the end of the experiment, the comet assay of blood and SOD, GSH-Px, MDA and 8-OHdG of liver were tested. The activities of SOD and GSH-Px in DiBP groups were significantly lower than the control group (P < 0.05); the MDA contents of DiBP group III and group IV were significantly higher than the control group (P < 0.05) and the 8-OHdG content of group II was significantly higher than the control group (P < 0.01). The comet assay showed that the oxidative damage of DNA in DiBP groups was significant in comparison with the control group (P < 0.05). Oxidative stress induced by diisobutyl phthalate can decrease the activities of antioxidative enzymes and result in oxidative damage of tissues.
Related Materials