EMAIL THIS PAGE TO A FRIEND
Free radical research
Reactions of reactive oxygen species (ROS) with curcumin analogues: Structure-activity relationship.
PMID 21034358
Abstract
Three curcumin analogues viz., bisdemethoxy curcumin, monodemethoxy curcumin, and dimethoxycurcumin that differ at the phenolic substitution were synthesized. These compounds have been subjected for free radical reactions with DPPH radicals, superoxide radicals (O(2)(•-)), singlet oxygen ((1)O(2)) and peroxyl radicals (CCl(3)O(2)(•)) and the bimolecular rate constants were determined. The DPPH radical reactions were followed by stopped-flow spectrometer, (1)O(2) reactions by transient luminescence spectrometer, and CCl(3)O(2)(•) reactions using pulse radiolysis technique. The rate constants indicate that the presence of o-methoxy phenolic OH increases its reactivity with DPPH and CCl(3)O(2)(•), while for molecules lacking phenolic OH, this reaction is very sluggish. Reaction of O(2)(•-) and (1)O(2) with curcumin analogues takes place preferably at β-diketone moiety. The studies thus suggested that both phenolic OH and the β-diketone moiety of curcumin are involved in neutralizing the free radicals and their relative scavenging ability depends on the nature of the free radicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90593
Demethoxycurcumin, analytical standard
C20H18O5
D7696
Demethoxycurcumin, ≥98% (HPLC)
C20H18O5