EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
Histamine: from flop to flip-flop.
PMID 21037309
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0593 CGP52432, ≥98% (HPLC)
C15H24Cl2NO4P