EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Cooperation between LepA and PlcH contributes to the in vivo virulence and growth of Pseudomonas aeruginosa in mice.
PMID 21041488
Abstract
Pseudomonas aeruginosa-derived large extracellular protease (LepA) and hemolytic phospholipase C (PlcH) are considered to play an important role in the pathogenicity of this organism. Although bacterial growth appears to be closely related to virulence, little is known about whether LepA and PlcH participate in the growth and virulence of P. aeruginosa. In this study, we investigated whether LepA and PlcH contribute to the virulence and growth of P. aeruginosa using a wild-type strain and mutants. The growth rate of the isogenic lepA single mutant was lower than that of the wild-type strain in a minimal medium containing serum albumin or hemoglobin as the sole carbon and nitrogen source. Furthermore, the growth rate of the lepA plcH double mutant decreased greatly compared with that of the wild-type strain in a minimal medium containing erythrocytes as a sole nutrient source for growth. Thus, these results indicate that cooperation between LepA and PlcH would contribute to the utilization of erythrocytes as a sole nutrient source for the growth of P. aeruginosa. In addition, mouse infection experiments demonstrated that the virulence of the lepA and plcH single mutants was attenuated, and the numbers of the mutants were lower than the numbers of the wild-type strain in peritoneal lavage fluid and whole-blood specimens. In particular, the virulence and growth rate of the lepA plcH double mutant were markedly lower than those of the wild-type strain. Collectively, these results suggest that LepA and PlcH contribute to the in vivo virulence and growth of P. aeruginosa.
Related Materials