EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section D, Biological crystallography
Using neutron protein crystallography to understand enzyme mechanisms.
PMID 21041947
Abstract
A description is given of the results of neutron diffraction studies of the structures of four different metal-ion complexes of deuterated D-xylose isomerase. These represent four stages in the progression of the biochemical catalytic action of this enzyme. Analyses of the structural changes observed between the various three-dimensional structures lead to some insight into the mechanism of action of this enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X4625
D-Xylulose, ≥98% (HPLC), syrup
C5H10O5