EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Validation of capillary electrophoresis method for determination of N-methylpyrrolidine in cefepime for injection.
PMID 21044414
Abstract
The present study relates to a new capillary electrophoresis method for the determination of N-methylpyrrolidine, an impurity considered to be toxic and also potential degradation impurity in cefepime hydrochloride drug substance. The newly developed capillary electrophoresis method for determining the content of N-methylpyrrolidine in cefepime for injection has been validated as per International Conference on Harmonization (ICH) guidelines to prove the selectivity, sensitivity, suitability, robustness, and ruggedness of the method. This simple, efficient, and rapid methodology may be used by pharmaceutical industry for routine analysis as well as during stability studies. The newly developed capillary electrophoresis method to determine the content of N-methylpyrrolidine in cefepime for injection requires 10 min for data acquisition, and uses an indirect UV photometry method to detect the analyte signal at 240 nm against the reference signal at 210 nm. The electrophoretic system is optimized to get stable base line, higher signal to noise ratio and peaks with narrow peak width. The method employs bare fused silica capillary with extended light path, effective length of capillary is 56 cm and inner diameter of capillary is 50 μm, 5 mmole of imidazole buffer adjusted to pH 5.1 with 3 molar acetic acid solution is used as background electrolyte. The sample is introduced in hydrodynamic mode employing pressure of 50 mbar for 5 s, and the desired separation is achieved with constant applied voltage of 25 kV at ambient temperature (~25°C).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69110
1-Methylpyrrolidine, ≥98.0% (GC)
C5H11N
M79204
N-Methylpyrrolidine, 97%
C5H11N