EMAIL THIS PAGE TO A FRIEND
Endocrine
Selective α1-adrenoceptor antagonist (controlled release tablets) in preoperative management of pheochromocytoma.
PMID 21046486
Abstract
The objective of this article is to evaluate the efficacy of Doxazosin Mesylate Controlled Release Tablets for preoperative treatment of patients with pheochromocytoma. Between 2003 and 2008, 67 patients with confirmed diagnoses of pheochromocytoma were enrolled in this study. According to the drug used in preoperative management, patients were divided into two groups: Doxazosin Mesylate pretreatment group (n=36) and Phenoxybenzamine pretreatment group (n=31). Surgery was performed only in patients who met the optimal preoperative condition. The hematocrit decreased significantly (P<0.001) after antiadrenergic therapy in patients pretreated with phenoxybenzamine or doxazosin. There was no significant difference between the fluid intakes during operation in both groups. The systolic arterial pressures both before and after induction of anesthesia were all significantly higher in the doxazosin patients than in the phenoxybenzamine group (P<0.05). After tumor removed, the lowest systolic arterial pressure was significantly higher in doxazosin group than in phenoxybenzamine group (P<0.05). The fluctuation of systolic arterial pressure during operation was more stable in doxazosin group than in phenoxybenzamine group (P<0.05). Doxazosin mesylate controlled release tablet was as effective as phenoxybenzamine in preoperative volume expansion. Although phenoxybenzamine provided better arterial pressure control, patients pretreated with DOX experienced more stable perioperative hemodynamic changes, shorter preoperative management periods and more simple medication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B019
Phenoxybenzamine hydrochloride, ≥97%, powder
C18H22ClNO · HCl