EMAIL THIS PAGE TO A FRIEND
Tissue & cell
Effects of some pesticides on the vital organs of juvenile rainbow trout (Oncorhynchus mykiss).
PMID 21051065
Abstract
Gill, trunk kidney, spleen, and liver of rainbow trout (Oncorhynchus mykiss) were examined after exposure to different sublethal concentrations of carbosulfan (25, 50 and 200 μgL(-1)), propineb (3, 6 and 24 mgL(-1)), and benomyl (2, 5 and 20 mgL(-1)) for 14 days. Lesions were observed in gill, trunk kidney, spleen, and liver of rainbow trout exposed to either concentration of pesticides. The most important lesions were determined in the highest concentrations of pesticides. Lamellar fusion, lamellar hyperplasia, epithelial lifting, vacuolization of epithelial tissue, epithelial necrosis, hypertrophy and sloughing of epithelium were observed on fish exposed to carbosulfan, propineb and benomyl. Fish had cell necrosis, degeneration and oedemas in liver, trunk kidney and spleen. None of these lesions were seen in control fish.
Related Materials