EMAIL THIS PAGE TO A FRIEND
Hormone research in paediatrics
Comments on 'Prospective study confirms oxandrolone-associated improvement in height in growth hormone-treated adolescent girls with Turner syndrome' by Zeger et al., pp. 39-47, this issue.
PMID 21051854
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0437
Oxandrolone, ≥98% (HPLC)
C19H30O3