EMAIL THIS PAGE TO A FRIEND
Addiction (Abingdon, England)
An anti-inflammatory as a recreational drug in Brazil.
PMID 21059186
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5524
Benzydamine hydrochloride, analytical standard
C19H23N3O · HCl