EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Pleasurable behaviors reduce stress via brain reward pathways.
PMID 21059919
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-519
Anti-phospho-CREB (Ser133) Antibody, clone, Upstate®, from rabbit