EMAIL THIS PAGE TO A FRIEND
Radiation protection dosimetry
Recalibration of indium foil for personnel screening in criticality accidents.
PMID 21062798
Abstract
At the Nuclear Fuel Cycle Engineering Laboratories of the Japan Atomic Energy Agency (JAEA), small pieces of indium foil incorporated into personal dosemeters have been used for personnel screening in criticality accidents. Irradiation tests of the badges were performed using the SILENE reactor to verify the calibration of the indium activation that had been made in the 1980s and to recalibrate them for simulated criticalities that would be the most likely to occur in the solution process line. In addition, Monte Carlo calculations of the indium activation using the badge model were also made to complement the spectral dependence. The results lead to a screening level of 15 kcpm being determined that corresponds to a total dose of 0.25 Gy, which is also applicable in posterior-anterior exposure. The recalibration based on the latest study will provide a sounder basis for the screening procedure in the event of a criticality accident.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF97000860
Indium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.999%
In
GF40799616
Indium, foil, 10mm disks, thickness 0.03mm, 99.8%
In
GF29634228
Indium, foil, 6mm disks, thickness 0.1mm, as rolled, 100%
In
GF74341278
Indium, foil, 6mm disks, thickness 0.20mm, as rolled, 100%
In
GF94704644
Indium, foil, 6mm disks, thickness 0.25mm, as rolled, 100%
In
GF83950027
Indium, foil, 6mm disks, thickness 0.5mm, as rolled, 100%
In
GF58678958
Indium, foil, 6mm disks, thickness 0.75mm, as rolled, 100%
In
GF44006712
Indium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
In
GF04295538
Indium, foil, 8mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%
In
GF87572649
Indium, foil, 8mm disks, thickness 0.015mm, 99.8%
In
GF85401918
Indium, foil, 8mm disks, thickness 0.01mm, 99.8%
In
GF40183950
Indium, foil, 8mm disks, thickness 0.025mm, 99.8%
In
GF89773138
Indium, foil, 8mm disks, thickness 0.02mm, 99.8%
In
GF76326396
Indium, foil, 8mm disks, thickness 0.03mm, 99.8%
In
GF06331047
Indium, foil, 8mm disks, thickness 0.04mm, 99.8%
In
GF35535157
Indium, foil, 8mm disks, thickness 0.05mm, as rolled, 100%
In
GF98837319
Indium, foil, 8mm disks, thickness 0.075mm, as rolled, 100%
In
GF39812137
Indium, foil, 8mm disks, thickness 0.125mm, as rolled, 100%
In
GF04277729
Indium, foil, 8mm disks, thickness 0.1mm, as rolled, 100%
In
GF92659540
Indium, foil, 8mm disks, thickness 0.20mm, as rolled, 100%
In
GF38080065
Indium, foil, 8mm disks, thickness 0.25mm, as rolled, 100%
In
GF93752590
Indium, foil, 8mm disks, thickness 0.5mm, as rolled, 100%
In
GF55033589
Indium, foil, 8mm disks, thickness 0.75mm, as rolled, 100%
In
GF00838997
Indium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%
In
GF24434748
Indium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 100%
In
GF61338966
Indium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 100%
In
GF29364981
Indium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 100%
In
GF07860900
Indium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 100%
In
GF96094873
Indium, foil, not light tested, 100x100mm, thickness 0.006mm, temporary acrylic support, 99.8%
In
GF97485823
Indium, foil, not light tested, 100x100mm, thickness 0.008mm, temporary acrylic support, 99.8%
In