EMAIL THIS PAGE TO A FRIEND
The Johns Hopkins medical letter health after 50
Consultation corner. The latest on female sexual health.
PMID 21066837
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0797 Flibanserin, ≥98% (HPLC)
C20H21F3N4O