EMAIL THIS PAGE TO A FRIEND
Mutagenesis
An overview of the visualisation and quantitation of low and high MW DNA adducts using the trapped in agarose DNA immunostaining (TARDIS) assay.
PMID 21068206
Abstract
The ability to detect and quantify specific DNA adducts benefits genome stability research, drug development and the evaluation of environmental mutagens. The trapped in agarose DNA immunostaining (TARDIS) assay was developed as a means of detecting and quantifying melphalan and cisplatin DNA adducts at the single-cell level and has since been adapted to quantify topoisomerase-DNA complexes. The method relies on salt-detergent extraction of agarose-embedded cells. Genomic DNA and any covalently attached molecules remain in place in the agarose, while other cellular constituents are removed. Drug-DNA or topoisomerase-DNA complexes are then detected and quantified by sensitive immunofluorescence using adduct-specific antibodies. Here, we give a perspective of the TARDIS assay including a comparison with other methods for quantifying topoisomerase-DNA covalent complexes and provide technical details required to set up and perform the assay.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6013
Agarose, Type I, low EEO
A6877
Agarose, Medium EEO, for molecular biology
A0169
Agarose, Type I-A, low EEO
A4718
Agarose, High resolution, for molecular biology
A4679
Agarose, Low EEO, for Immunoelectrophoresis
A7431
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
A9918
Agarose, Type II-A, Medium EEO
A0576
Agarose, Low EEO
A2790
Agarose, Wide range, for molecular biology
A9311
Agarose, Medium EEO, for electrophoresis
A3893
Agarose, Type VI, High Gelling Temperature
A2929
Agarose, For pulsed field electrophoresis running gel
A3038
Agarose, Type XI, low gelling temperature, suitable for separation of small nucleic acid fragments
A5030
Agarose, Ultra-low Gelling Temperature
A3643
Agarose, Type IV, Special High EEO
A2576
Agarose, Ultra-low Gelling Temperature, molecular biology grade
A9793
Agarose, High EEO
A4905
Agarose, Type VIII
A7174
Agarose, High Gelling Temperature
A5304
Agarose, Special High EEO
A3054
Agarose, For pulsed field electrophoresis sample preparation
05066
Agarose, High EEO, for molecular biology
4B200
Sepharose® 4B, 45-165 μm bead diameter
GE17-0120-01
Sepharose® 4B, GE Healthcare, 17-0120-01, pack of 1 L
6B100
Sepharose® 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
GE17-0110-01
Sepharose® 6B, GE Healthcare, 17-0110-01, pack of 1 L
U0507
Ultrogel® A4, aqueous ethanol suspension, 60-140 μm (wet bead diameter), exclusion limit 20,000,000 Da, fractionation range 55,000-9,000,000 Da (globular proteins)