EMAIL THIS PAGE TO A FRIEND
Bioscience Reports
A brain-derived MeCP2 complex supports a role for MeCP2 in RNA processing.
PMID 21070191
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-013
Anti-MeCP2 Antibody, clone, Upstate®, from rabbit