EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Anticholinesterase activity of standardized extract of Illicium verum Hook. f. fruits.
PMID 21075180
Abstract
Illicium verum is a well known spice in traditional Indian system for its therapeutic potential. The present study was aimed to evaluate the acetylcholinesterase (AChE) and butyrylcholinesterase inhibitory (BChE) activity of standardized extracts of I. verum and its oil. Present study confirmed that anethole contributed to the anticholinesterase activity of I. verum, with more specificity towards AChE. IC(50) for AChE and BChE inhibitory activity of anethole was 39.89±0.32 μg/mL and 75.35±1.47 μg/mL, whereas for the oil, 36.00±0.44 μg/mL and 70.65±0.96 μg/mL respectively. Therefore I. verum can be a good lead as anti-cholinesterase agent from natural resources.