EMAIL THIS PAGE TO A FRIEND
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde
Combination of a thiazide, a vasodilator and reserpine compared with methyldopa plus hydrochlorothiazide in the treatment of hypertension in Zimbabwe.
PMID 2107581
Abstract
Brinerdin (Sandoz), a combination of a diuretic (clopamide 5 mg), a vasodilator (dihydro-ergocristine 0.5 mg) and reserpine (0.1 mg) (CDR) was compared with methyldopa (MD) plus hydrochlorothiazide (HCT) for antihypertensive effect, adverse reactions, compliance and patient preference in an open cross-over trial. Eighteen patients completed both arms of the trial and 5 patients who completed the CDR arm were withdrawn while on the MD arm because of adverse effects in 4 and poor control in 1. On HCT 50 mg daily the mean baseline systolic blood pressure was 163.9 +/- 16.3 mmHg and the diastolic blood pressure was 105.9 +/- 6.7 mmHg. On CDR these were reduced to systolic blood pressure 140.3 +/- 15.1 mmHg and diastolic blood pressure 87.8 +/- 9.3 mmHg. On MD + HCT the systolic blood pressure was reduced to 138.5 +/- 16.9 mmHg and the diastolic blood pressure to 88.9 +/- 10.3 mmHg. The differences between the two treatment periods in systolic blood pressure (1.8 mmHg; 95% confidence interval (CI) - 4.1 + 7.7 mmHg) and diastolic blood pressure (1.1 mmHg; 95% CI - 4.6 + 2.4 mmHg) were not significant with P values of 0.6 and 0.7 respectively. Compliance was 98.2% for CDR and 94.7% for MD + HCT (P = 0.02). Unusual sleepiness occurred more frequently in the MD arm (P less than 0.01). Thirteen patients chose to continue on CDR, 2 on MD + HCT and 3 had no preference (P = 0.005). CDR is similar in antihypertensive effect to MD + HCT but is better tolerated with fewer withdrawals, fewer adverse effects, better compliance and has more patients electing to continue taking it.