EMAIL THIS PAGE TO A FRIEND
Clinical and experimental obstetrics & gynecology
The slow levonorgestrel-releasing intrauterine system (LNG-IUS) 20 mcg/day: a literature review.
PMID 21077492
Abstract
The aim of the study is to present the mechanisms of action, indications, complications, contraindications and the necessary tests before the insertion of a levonorgestrel-releasing intrauterine system (Mirena). After a literature search in Pubmed, a narrative review in the field is presented.