EMAIL THIS PAGE TO A FRIEND
Cephalalgia : an international journal of headache
Efficacy and tolerability of rizatriptan for the treatment of acute migraine in sumatriptan non-responders.
PMID 21078681
Abstract
The study was carried out to assess the efficacy and tolerability of rizatriptan orally disintegrating tablet (ODT) for treating acute migraine in patients who are non-responders to sumatriptan. Many migraineurs report dissatisfaction with sumatriptan efficacy. It is unclear whether sumatriptan 100 mg non-responders will respond to other triptans. This was a randomized, placebo-controlled, double-blind study in adults with >1-year history of ICHD-II (International Classification of Headache Disorders, second edition) migraine who reported that they generally do not respond to sumatriptan (≥50% unsatisfactory response). In the baseline phase, participants treated a single moderate/severe migraine attack with open-label generic sumatriptan 100 mg. Those who continued to experience moderate/severe pain at two hours post-dose were eligible to enter the double-blind treatment phase, during which participants treated three migraine attacks in crossover fashion (two with rizatriptan 10-mg ODT, one with placebo) after being randomly assigned to one of three treatment sequences (1 : 1 : 1 ratio). The primary endpoint was two-hour pain relief. A total of 102 (94%) participants treated at least one study migraine. Pain relief at two hours was significantly greater with rizatriptan compared with placebo (51% vs. 20%, p < .001). Response rates also favored rizatriptan on two-hour pain freedom (22% vs. 12%, p = .013) as well as 24-hour sustained pain relief (38% vs. 14%, p < .001) and sustained pain freedom (20% vs. 11%, p = .036). Treatment was generally well tolerated. Rizatriptan 10-mg ODT was superior to placebo at providing two-hour pain relief and two-hour pain freedom in the treatment of acute migraine in those who do not respond to sumatriptan 100 mg. Rizatriptan was generally well tolerated in this population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0247
Rizatriptan benzoate salt, ≥98% (HPLC)
C15H19N5 · C7H6O2