EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Extended para-hydrogenation monitored by NMR spectroscopy.
PMID 21079839
Abstract
A system that provides a sustained hyperpolarized (1)H NMR signal in an aqueous medium is reported. The enhanced signal lasts much longer than typical (1)H T(1) values, uncovering new possibilities for implementing hyperpolarized (1)H NMR/MRI experiments or performing kinetics studies that would not otherwise be detectable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317519
Methyl 2-acetamidoacrylate, 98%
C6H9NO3