EMAIL THIS PAGE TO A FRIEND
Amino acids
A practical synthesis of N α-Fmoc protected L-threo-β-hydroxyaspartic acid derivatives for coupling via α- or β-carboxylic group.
PMID 21082204
Abstract
A simple and practical general synthetic protocol towards orthogonally protected tHyAsp derivatives fully compatible with Fmoc solid-phase peptide synthetic methodology is reported. Our approach includes enantioresolution of commercially available D: ,L: -tHyAsp racemic mixture by co-crystallization with L: -Lys, followed by ion exchange chromatography yielding enantiomerically pure L: -tHyAsp and D: -tHyAsp, and their selective orthogonal protection. In this way N ( α )-Fmoc protected tHyAsp derivatives were prepared ready for couplings via either α- or β-carboxylic group onto the resins or the growing peptide chain. In addition, coupling of tHyAsp via β-carboxylic group onto amino resins allows preparation of peptides containing tHyAsn sequences, further increasing the synthetic utility of prepared tHyAsp derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2775
DL-threo-β-Hydroxyaspartic acid
C4H7NO5