EMAIL THIS PAGE TO A FRIEND
International journal of oral and maxillofacial surgery
Miniplate osteosynthesis with four different systems in sheep.
PMID 21084173
Abstract
The aim of this study was to compare a combination of a locking system with self-tapping (ST-L) or self-drilling-tapping (SDT-L) screws with a combination of conventional miniplates with self-tapping (ST) and self-forming (SF) screws. A standardized osteotomy and osteosynthesis with one of the above mentioned systems was performed in 24 sheep. Callus formation was measured with the help of CT scans assisted by a navigation system. Specimens of each osteotomy gap were taken and examined histologically. The best results were observed when self-tapping screws and the Mini-Locking-System (ST-L) were applied. The slowest healing was seen in animals treated with miniplates and SF screws. After 8 weeks an increase in bone formation could be observed in the ST, SF, SDT-L systems. The results after 8 weeks were comparable with those achieved by the ST-L system after 4 weeks. The improved stability of the osteosynthesis with the ST-L system resulted in early ossification of the osteotomy gap and the smallest amount of callus formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52097
Xylenol Orange disodium salt, for spectrophotometric determination of metal ions
C31H30N2Na2O13S