EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
A computer-automated touchscreen paired-associates learning (PAL) task for mice: impairments following administration of scopolamine or dicyclomine and improvements following donepezil.
PMID 21086119
Abstract
Performance on the Cambridge Neuropsychological Test Automated Battery touchscreen paired-associates learning (PAL) test is predictive of Alzheimer's disease and impaired in schizophrenia and chronic drug users. An automated computer touchscreen PAL task for rats has been previously established. A pharmacologically validated PAL task for mice would be a highly valuable tool, which could be useful for a number of experimental aims including drug discovery. This study sought to investigate the effects of systemic administration of cholinergic agents on task performance in C57Bl/6 mice. Scopolamine hydrobromide (0.02, 0.2, and 2.0 mg/kg), dicyclomine hydrochloride (M(1) receptor antagonist; 2.0, 4.0, and 8.0 mg/kg), and donepezil hydrochloride (cholinesterase inhibitor; 0.03, 0.1, and 0.3 mg/kg) were administered post-acquisition in C57Bl/6 mice performing the PAL task. Scopolamine (0.2 and 2.0 mg/kg) and dicyclomine (at all administered doses) significantly impaired PAL performance. A significant facilitation in PAL was revealed in mice following donepezil administration (0.3 mg/kg). The present study shows that mice can acquire the rodent PAL task and that the cholinergic system is important for PAL task performance. M(1) receptors in particular are likely implicated in normal performance of PAL. The finding that mouse PAL can detect both impairments and improvements indicates that this task could prove to be a highly valuable tool for a number of experimental aims including drug discovery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7909
Dicyclomine hydrochloride, ≥99% (TLC), powder
C19H35NO2 · HCl