EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Enantiospecific synthesis of 2-[18F]fluoro-L-phenylalanine and 2-[18F]fluoro-L-tyrosine by isotopic exchange.
PMID 21088800
Abstract
2-[(18)F]Fluoro-L-phenylalanine and 2-[(18)F]fluoro-L-tyrosine have been developed as promising radiopharmaceuticals for molecular imaging using positron emission tomography (PET). However, the lack of a convenient radiosynthetic pathway has limited their practical use. In this work a new three-step nucleophilic synthesis of these compounds starting from [(18)F]fluoride is described. Corresponding precursors (1a and 1b) were (18)F-fluorinated by isotopic exchange, followed by the removal of an activating formyl group with Rh(PPh(3))(3)Cl and subsequent hydrolysis of protecting groups in acidic medium. All reactions were carried out using both conventional and microwave heating. Conventional heated reactions yielded the desired products 2-[(18)F]Fphe and 2-[(18)F]Ftyr in 43% and 49% whereas radiochemical yields of 34% and 43%, respectively, were obtained when they were heated by microwaves. Under optimized conditions the enantiomeric purity was ≥94% for both radiopharmaceuticals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47300
o-Fluoro-DL-phenylalanine, ≥98%
C9H10FNO2