EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[Development of anti-trypanosome drugs targeting nucleotides biosynthesis and red-ox regulatory pathway].
PMID 21089606
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
537306
D-Hydroorotic acid, 98%
C5H6N2O4
D7003
DL-Dihydroorotic acid
C5H6N2O4
D7128
L-Dihydroorotic acid, ≥99%
C5H6N2O4