EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
2-Nitropropane.
PMID 21089925
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130265
2-Nitropropane, ≥96%
C3H7NO2