EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
N-nitrosodiethanolamine.
PMID 21089927
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7632
N-Nitrosodiethanolamine
C4H10N2O3