EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of methyl N-Boc-(2S,4R)-4-methylpipecolate.
PMID 21090804
Abstract
An efficient stereoselective synthesis of fully protected (2S,4R)-4-methylpipecolic acid has been developed. The synthesis was achieved by initial asymmetric α-alkylation of glycine with a chiral iodide, affording the linear precursor as a single stereoisomer. Subsequent aldehyde formation using OsO(4)/NaIO(4) followed by immediate intramolecular cyclization afforded an enamine that was then subjected to hydrogenation to give the final compound in 23% yield over 10 steps.