EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
A new and efficient synthetic route for the anxiolytic agent CL285032.
PMID 21095124
Abstract
CL285032 is an anxiolytic compound currently under investigation as a possible treatment for canine noise phobia associated anxiety. A robust scale-up and manufacturing process is essential for the development and marketability of the drug. The current synthetic route, although reliable, requires seven steps and has a low overall yield (18%), leaving opportunity for improvement. We are presenting an efficient alternative approach toward the synthesis of CL285032 and the results thereof.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P57204
Pyridazine, 98%
C4H4N2