EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A dual host approach to transmembrane transport of salts.
PMID 21103515
Abstract
A dual host approach for M(+)/Cl(-) co-transport has been shown to be effective in lipid bilayers consisting of POPC using fluorescence-based transport assays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8010
N,N′-Dimethyl-9,9′-biacridinium dinitrate, used as chemiluminescent reagent
C28H22N4O6