EMAIL THIS PAGE TO A FRIEND
Natural product research
A tetrahydrofuranol from the leaves of Michelia compressa var. lanyuensis (Magnoliaceae).
PMID 21104528
Abstract
Pressalanin A, a 2-functionalized tetrahydrofuranol, has been isolated from the leaves of Michelia compressa var. lanyuensis (Magnoliaceae), and the structure was determined on the basis of a spectroscopic analysis.