EMAIL THIS PAGE TO A FRIEND
Natural product research
Composition of essential oils in subterranean organs of three species of Valeriana L.
PMID 21104529
Abstract
Essential oils from the subterranean organs of three species of Valeriana L. from Iran (Valeriana sisymbriifolia Vahl, Valeriana alliariifolia Adams and Valeriana officinalis L.) belonging to Valerianaceae family have been obtained by hydrodistillation and analysed by gas chromatography-mass spectrometry in order to discern the differences and similarities between the volatile chemical compositions of these species. More than 100 components were identified in essential oils of the studied plants (Supplementary Table S1--online only). The principal common constituents of the three species of Valeriana were spathulenol, limonene, γ-terpinene, vulgarone B and p-cymene. The main essential oil ingredients were α-selinene (7.83%) in V. sisymbriifolia, limonene (3.53%) in V. alliariifolia and spathulenol (13.33%), α-campholenal (11.48%), vulgarone B (8.38%) and valerenal (8.32%) in V. officinalis plants. Ageratochromene (precocene II), a chromene substance with antibacterial, antifungal, insecticidal and antijuvenile hormonal activities, was found at high levels (35.59% and 36.58%) in the essential oils of V. sisymbriifolia plants.