EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Michler's ketone (4,4'-(dimethylamino)benzophenone).
PMID 21105249
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
147834
Michler’s ketone, 98%
C17H20N2O