EMAIL THIS PAGE TO A FRIEND
Journal of Neurochemistry
Climbing fiber-evoked Purkinje cell discharge reduces expression of GABA(A) receptor-associated protein and decreases its interaction with GABA(A) receptors.
PMID 21105873