EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A graphene oxide-based nano-beacon for DNA phosphorylation analysis.
PMID 21107474
Abstract
We designed a single-fluorophore-tagged hairpin-structured nano-beacon probe by using a superquencher, graphene oxide (GO), based on which a new method for the analysis of DNA phosphorylation detection was developed.