EMAIL THIS PAGE TO A FRIEND
Metabolic Brain Disease
Glutamine synthetase gene expression and glutamate transporters in C6-glioma cells.
PMID 21107897
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1783
Anti-Glutamate Transporter Antibody, Glial, serum, clone, Chemicon®