EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Comment on "A glycerol-free process to produce biodiesel by supercritical methyl acetate technology: an optimization study via response surface methodology".
PMID 21109431