EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Enzymatic synthesis of (125/131)I labeled 8-hydroxyquinoline glucuronide and in vitro/in vivo evaluation of biological influence.
PMID 21109446
Abstract
8-Hydroxyquinoline (8-OHQ) is a long-known molecule which due to its metal-complexation ability is frequently used for analysis. It is also called oxine. Oxine and derivatives have been investigated to process antitumor and antimicrobial activities. 8-Hydroxyquinolyl-glucuronide (8-OHQ-Glu) was enzymatically synthesized using microsome preparates separated from Hutu-80 cells, labeled with (125)I to perform a radionuclide labeled prodrug and investigated of its biological affinities on Hutu-80 (human duodenum intestinal adenocarcinoma), Caco-2 (human colorectal adenocarcinoma), Detroit 562 (human pharynx adenocarcinoma) cells and ACBRI 519 (primary human small intestine epithelial cells) in this work. UDP-glucuronyl transferase (UDPGT) rich microsome preparates, which are used for glucuronidation in enzymatic synthesis, were extracted from Hutu-80 cells. 8-OHQ-Glu components were labeled using iodogen method with (125)I and (131)I. Structural analyses were performed with LC/MS/MS, (1)H NMR and (13)C-MMR for identify and measure chemical constituents. Results confirmed expected molecular structure. 8-OHQ-Glu could successfully radioiodinated with (125/131)I according to iodogen method. (125)I-8-OHQ-glucuronide incorporated with human gastrointestinal cancer cells such as Detroit-562 (human pharynx adenocarcinoma) (12.6%), Caco-2 (human colorectal adenocarcinoma) (7.8%), Hutu- 80 (human duodenum intestinal adenocarcinoma) (9.5%) and ACBRI 519 (primary human small intestine epithelial cells) (6.40%). (131)I-8-OHQ-Glu was tested in mice bearing subcutaneously implanted Caco-2 colorectal adenocarcinoma cells. The results demonstrated that radioiodinated 8-OHQ-Glu may be promising anticancer prodrug.