EMAIL THIS PAGE TO A FRIEND
Talanta
A new colorimetric chemodosimeter for Hg2+ based on charge-transfer compound of N-methylpyrrole with TCNQ.
PMID 21111186
Abstract
Reaction of N-methylpyrrole and 7,7,8,8-tetracyanoquinodimethane (TCNQ) furnishes an intense blue unsymmetrical charge-transfer compound through regioselective attachment of tricyanoquinodimethane at the 2-position of N-methylpyrrole which was found to be selective chemodosimeter for Hg(2+) ions in CH(3)CN:H(2)O mixture (1:1 v/v, pH=7.0, 0.01 M HEPES, 0.15M NaCl) as well as in the solid state when supported on silica, over a variety of metal ions. A plausible mechanism for the sensing process has been proposed and supported through the characterization of the resulting Hg(2+) complex and the density functional theory (DFT) studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W509973
1-Methylpyrrole, ≥99%
C5H7N
M78801
N-Methylpyrrole, 99%
C5H7N