EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Anti-inflammatory effect of visnagin in lipopolysaccharide-stimulated BV-2 microglial cells.
PMID 21116788
Abstract
Visnagin, which is found in Ammi visnaga, has biological activity as a vasodilator and reduces blood pressure by inhibiting calcium influx into the cell. The present study demonstrates the anti-inflammatory effect of visnagin on lipopolysaccharide (LPS)-stimulated BV-2 microglial cells. When cells were treated with visnagin prior to LPS stimulation, production of nitric oxide and expression of iNOS were attenuated in a dose-dependent manner. Visnagin also caused a significant decrease of mRNA expression and release of TNF-α, IL-1β and IFNγ. In addition, visnagin reduced LPS-induced IL-6 and MCP-1 mRNA level. We further found that visnagin dose-dependently inhibited LPS-induced AP-1 and NF-κB luciferase activities. Taken together, our results for the first time suggest that the anti-inflammatory effect of visnagin might result from the inhibition of transcription factors, such as AP-1 and NF-κB.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60730
Khellin, for microscopy
C14H12O5