EMAIL THIS PAGE TO A FRIEND
Combinatorial chemistry & high throughput screening
Parallel solution-phase synthesis of a 2-aminothiazole library including fully automated work-up.
PMID 21118081
Abstract
A straightforward and effective procedure for the solution phase preparation of a 2-aminothiazole combinatorial library is described. Reaction, work-up and isolation of the title compounds as free bases was accomplished in a fully automated fashion using the Chemspeed ASW 2000 automated synthesizer. The compounds were obtained in good yields and excellent purities without any further purification procedure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123129
2-Aminothiazole, 97%
C3H4N2S