EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Purification, characterization, and cloning of a bifunctional molybdoenzyme with hydratase and alcohol dehydrogenase activity.
PMID 21120472
Abstract
A bifunctional hydratase/alcohol dehydrogenase was isolated from the cyclohexanol degrading bacterium Alicycliphilus denitrificans DSMZ 14773. The enzyme catalyzes the addition of water to α,β-unsaturated carbonyl compounds and the subsequent alcohol oxidation. The purified enzyme showed three subunits in SDS gel, and the gene sequence revealed that this enzyme belongs to the molybdopterin binding oxidoreductase family containing molybdopterins, FAD, and iron-sulfur clusters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
481718
Iron(0) pentacarbonyl, >99.99% trace metals basis
C5FeO5
195731
Iron(0) pentacarbonyl
C5FeO5